Merck
  • Structure-activity relationships of boronic acid inhibitors of dipeptidyl peptidase IV. 1. Variation of the P2 position of Xaa-boroPro dipeptides.
Structure-activity relationships of boronic acid inhibitors of dipeptidyl peptidase IV. 1. Variation of the P2 position of Xaa-boroPro dipeptides.
Journal of medicinal chemistry (1996-05-10)
S J Coutts, T A Kelly, R J Snow, C A Kennedy, R W Barton, J Adams, D A Krolikowski, D M Freeman, S J Campbell, J F Ksiazek, W W Bachovchin
ABSTRACT
A series of prolineboronic acid (boroPro) containing dipeptides were synthesized and assayed for their ability to inhibit the serine protease dipeptidyl peptidase IV (DPPIV). Inhibitory activity, which requires the (R)-stereoisomer of boroPro in the P1 position, appears to tolerate a variety of L-amino acids in the P2 position. Substitution at the P2 position which is not tolerated include the D-amino acids, alpha,alpha-disubstituted amino acids, and glycine. Specificity against DPPII and proline specific endopeptidase is reported. A correlation between the ability to inhibit DPPIV in cell culture and in the human mixed lymphocyte reaction is demonstrated. A synthesis of prolineboronic acid is reported as well as conditions for generating the fully unprotected boronic acid dipeptides in either their cyclic or acyclic forms.
Sigma-Aldrich
Val-boroPro