Merck
  • Estrogen-responsive finger protein as a new potential biomarker for breast cancer.
Estrogen-responsive finger protein as a new potential biomarker for breast cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2005-09-08)
Takashi Suzuki, Tomohiko Urano, Tohru Tsukui, Kuniko Horie-Inoue, Takuya Moriya, Takanori Ishida, Masami Muramatsu, Yasuyoshi Ouchi, Hironobu Sasano, Satoshi Inoue
ABSTRACT
Estrogen-responsive finger protein (Efp) is a member of RING finger-B box-Coiled Coil family and is also a downstream target of estrogen receptor alpha. Previously, Efp was shown to mediate estrogen-induced cell growth, which suggests possible involvement in the development of human breast carcinomas. In this study, we examined expression of Efp in breast carcinoma tissues and correlated these findings with various clinicopathologic variables. Thirty frozen specimens of breast carcinomas were used for immunohistochemistry and laser capture microdissection/real-time PCR of Efp. Immunohistochemistry for Efp was also done in 151 breast carcinoma specimens fixed with formalin and embedded in paraffin wax. Efp immunoreactivity was detected in breast carcinoma cells and was significantly associated with the mRNA level (n = 30). Efp immunoreactivity was positively associated with lymph node status or estrogen receptor alpha status and negatively correlated with histologic grade or 14-3-3sigma immunoreactivity (n = 151). Moreover, Efp immunoreactivity was significantly correlated with poor prognosis of breast cancer patients, and multivariate analyses of disease-free survival and overall survival for 151 breast cancer patients showed that Efp immunoreactivity was the independent marker. Our data suggest that Efp immunoreactivity is a significant prognostic factor in breast cancer patients. These findings may account for an oncogenic role of Efp in the tumor progression of breast carcinoma.