Merck
  • Surface plasmon resonance (SPR) analysis of binding interactions of proteins in inner-ear sensory epithelia.
Surface plasmon resonance (SPR) analysis of binding interactions of proteins in inner-ear sensory epithelia.
Methods in molecular biology (Clifton, N.J.) (2008-10-08)
Dennis G Drescher, Neeliyath A Ramakrishnan, Marian J Drescher
ABSTRACT
Surface plasmon resonance is an optical technique utilized for detecting molecular interactions. Binding of a mobile molecule (analyte) to a molecule immobilized on a thin metal film (ligand) changes the refractive index of the film. The angle of extinction of light, reflected after polarized light impinges upon the film, is altered, monitored as a change in detector position for the dip in reflected intensity (the surface plasmon resonance phenomenon). Because the method strictly detects mass, there is no need to label the interacting components, thus eliminating possible changes of their molecular properties. We have utilized surface plasmon resonance to study the interaction of proteins of hair cells.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder