포장
pkg of 5 ea
1 of 4
이 품목 | HS120728 | HS120729 | HS120538 |
---|---|---|---|
manufacturer/tradename Heathrow Scientific 120727 | manufacturer/tradename Heathrow Scientific 120728 | manufacturer/tradename Heathrow Scientific 120729 | manufacturer/tradename Heathrow Scientific HS120538 |
material pink-purple , polypropylene , polycarbonate lid | material blue , polycarbonate lid, polypropylene | material blue , pink-purple , polycarbonate lid, polypropylene | material polypropylene |
packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pkg of 2 ea | packaging pkg of 5 |
feature holds 96-Well, 0.2 mL and 0.5 mL Tubes, and Assay Plates | feature holds 96-Well, 0.2 mL and 0.5 mL Tubes, and Assay Plates | feature 96-Well, 0.2 mL and 0.5 mL Tubes, and Assay Plates | feature Holds 96 x 0.2 mL tubes (individual or strips) |
L × W × H 110 mm × 148 mm × 48 mm | L × W × H 110 mm × 148 mm × 48 mm | L × W × H 110 mm × 148 mm × 48 mm | L × W × H - |
가장 최신 버전 중 하나를 선택하세요:
이미 열람한 고객
Karin E Koopman et al.
Hepatology (Baltimore, Md.), 60(2), 545-553 (2014-03-29)
American children consume up to 27% of calories from high-fat and high-sugar snacks. Both sugar and fat consumption have been implicated as a cause of hepatic steatosis and obesity but the effect of meal pattern is largely understudied. We hypothesized
Marieke G N Bos et al.
Psychoneuroendocrinology, 46, 102-113 (2014-06-03)
Retrieval of negative emotional memories is often accompanied by the experience of stress. Upon retrieval, a memory trace can temporarily return into a labile state, where it is vulnerable to change. An unresolved question is whether post-retrieval stress may affect
Nirit Geva et al.
Pain, 155(11), 2418-2425 (2014-09-25)
Anecdotes on the ability of individuals to continue to function under stressful conditions despite injuries causing excruciating pain suggest that acute stress may induce analgesia. However, studies exploring the effect of acute experimental stress on pain perception show inconsistent results
Daniëlle A I Groffen et al.
Social science & medicine (1982), 127, 143-149 (2014-10-14)
Lower educational attainment generally is a strong predictor of coronary heart disease (CHD). The underlying mechanisms of this effect are, however, less clear. One hypothesis is that stress related to limitations imposed by lower socioeconomic status elicits changes in hypothalamic-pituitary-adrenal
Lorenz Goette et al.
Psychoneuroendocrinology, 54, 115-123 (2015-02-24)
Social competition is a fundamental mechanism of evolution and plays a central role in structuring individual interactions and communities. Little is known about the factors that affect individuals' competitive success, particularly in humans. Key factors might include stress, a major