Product Name
3-Chloropropionyl chloride technical grade
Product Number
147443
제품 브랜드
ALDRICH
CAS Number
625-36-5
Molecular Weight
126.97
테스트 | 규격 |
---|---|
Appearance (Color) | Conforms to Requirements |
Colorless to Dark Brown | |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Purity (Titration by NAOH) | ≤105.0 % |
GC (area %) | ≥95.0 % |
VPCO |