Product Name
Fluoride on polymer support macroporous, 20-50 mesh, extent of labeling: 2.0-3.0 mmol/g loading
Product Number
387789
제품 브랜드
ALDRICH
CAS Number
39339-85-0
Molecular Weight
테스트 | 규격 |
---|---|
APPEARANCE | WHITE TO LIGHT BROWN OR LIGHT VIOLET SMALL |
BEADS | |
MISCELLANEOUS ASSAYS | REPORT MMOL F/G RESIN (~ 3%) |
TITRATION | 15-20% H2O |
APPROVED OCTOBER 20, 2008 RJM |