Product Name
2′,4′-Dimethylacetanilide 98%
Product Number
422894
제품 브랜드
ALDRICH
CAS Number
2050-43-3
Molecular Weight
163.22
테스트 | 규격 |
---|---|
Appearance (Color) | White |
Appearance (Form) | Conforms to Requirement |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Carbon | 71.7 - 75.4 % |
Nitrogen | 8.3 - 8.8 % |
Purity (GC) | ≥97.5 % |