Product Name
Formaldehyde dimethyl acetal for Grignard reactions, ≥99.0% (GC)
Product Number
47649
제품 브랜드
ALDRICH
CAS Number
109-87-5
Molecular Weight
76.09
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.0 % |
WATER (COULOMETR.) | ≤ 0.2 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |