Product Name
Hexadecylpyridinium bromide ≥97.0%
Product Number
52340
제품 브랜드
ALDRICH
CAS Number
140-72-7
Molecular Weight
384.44
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder |
ARGENTOMETR. TITRATION | Dry Substance |
WATER | FOR INFORMATION |
CARBON CONTENT | 62.67 % (THEORY) |
HYDROGEN CONTENT | 10.02 % (THEORY) |
NITROGEN CONTENT | 3.48 % (THEORY) |