Product Name
3-Phenylbenzyl bromide 97%
Product Number
538388
제품 브랜드
ALDRICH
CAS Number
14704-31-5
Molecular Weight
247.13
테스트 | 규격 |
---|---|
APPEARANCE | WHITE TO TAN POWDER, CRYSTALS OR CRYSTALLINE |
POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5% - 103.5% (WITH AGNO3 AFTER OXYGEN |
COMBUSTION) | |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED DECEMBER 5, 2005 BTR |