Product Name
Di(1-adamantyl)-n-butylphosphine hydriodide 95%
Product Number
671584
제품 브랜드
ALDRICH
CAS Number
714951-87-8
Molecular Weight
486.45
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
PHOSPHORUS 31 NMR SPECTRUM | CONFORMS TO STRUCTURE |
REMARKS | Chemical Purity > 95 % |
RECOMMENDED RETEST PERIOD | 40 MONTHS |