Product Name
(+)-Cinchonine 85%
Product Number
857270
제품 브랜드
ALDRICH
CAS Number
118-10-5
Molecular Weight
294.39
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder |
TITRATION (NT) HCLO4 0.1M | 84.5 - 115.5 % |
PURITY (HPLC AREA %) | ≥ 84.5 % |
OPTICAL ROTATION | 220.0 - 230.0 Degrees |
CONCENTRATION | C=0.5 IN ETOH |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |