Product Name
2-Amino-1-phenylethanol 98%
Product Number
A72405
제품 브랜드
ALDRICH
CAS Number
7568-93-6
Molecular Weight
137.18
테스트 | 규격 |
---|---|
APPEARANCE | WHITE POWDER, SOLID, CRYSTALS AND/OR CHUNKS |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 97.5%-102.5% (WITH HCLO4) |
THIN-LAYER | CONSISTENT WITH 98% PURITY |
SOLUBILITY | 50MG/ML, ACETIC ACID; CLEAR TO SLIGHTLY HAZY |
COLORLESS TO PALE YELLOW SOLUTION | |
REVISED OCTOBER 26, 2006 RJM |