Product Name
9-Phenylcarbazole 97%
Product Number
P21501
제품 브랜드
ALDRICH
CAS Number
1150-62-5
Molecular Weight
243.30
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | White to Light Yellow |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (GC AREA %) | ≥ 96.5 % |
CARBON CONTENT | 85.8 - 92.0 % |
NITROGEN CONTENT | 5.5 - 6.1 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |