Product Name
TEPP PESTANAL®, analytical standard
Product Number
32434
제품 브랜드
SIAL
CAS Number
107-49-3
Molecular Weight
290.19
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow to Light Brown-Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 98.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
RECOMMENDED RETEST PERIOD | 72 MONTHS |