Product Name
Tetratriacontane analytical standard
Product Number
88152
제품 브랜드
SIAL
CAS Number
14167-59-0
Molecular Weight
478.92
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | Colorless or White |
APPEARANCE (FORM) | Powder or Crystals or Flakes |
PURITY (GC AREA %) | ≥ 98.0 % |
MELTING POINT | 72 - 75 C |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |