Product Name
Activated Charcoal Norit® Norit® RB3, steam activated, rod
Product Number
29204
제품 브랜드
SIGALD
CAS Number
7440-44-0
Molecular Weight
12.01
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | Black |
APPEARANCE (FORM) | Rods |
PARTICLE SIZE | diam. ~3mm |
LOSS ON DRYING | ≤ 5 % |
APPLICATION TEST | butane adsorption 18-23 % p/p0 = 1 |