Product Name
Atto 655 NHS ester BioReagent, suitable for fluorescence
Product Number
76245
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
APPEARANCE (COLOR) | Report Result |
APPEARANCE (FORM) | Report Result |
PURITY (HPLC AREA %) | ≥ 90.0 % |
Absorption Max | 650.0 - 656.0 nm |
SOLVENT (UV) | ethanol with 0.1% trifluoroacetic acid |
DEGREE OF COUPLING | ≥ 90.0 % |
RECOMMENDED RETEST PERIOD | 48 MONTHS |