Product Name
Monoclonal Anti-SNAI1 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL3700, purified immunoglobulin, buffered aqueous glycerol solution
Product Number
AMAB91215
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Immunohistochemistry Dilution | 1:5000 - 1:10000 |
Immunohistochemistry Antibody Retrieval Method | HIER pH6 |