Product Name
Cantharidin
Product Number
C7632
제품 브랜드
SIGMA
CAS Number
56-25-7
Molecular Weight
196.20
테스트 | 규격 |
---|---|
APPEARANCE | WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS TO VERY FAINT YELLOW SOLUTION AT 25MG/ML IN ACETONE |
ELEMENTAL ANALYSIS | 60.0 TO 62.4% CARBON |
PURITY BY GAS CHROMATOGRAPHY | NLT 98% |
DOCUMENT # C7632/12/23/97/2 |