Product Name
Gly-Pro 4-methoxy-β-naphthylamide dipeptidylpeptidase IV substrate
Product Number
G9137
제품 브랜드
SIGMA
CAS Number
42761-76-2
Molecular Weight
327.38
테스트 | 규격 |
---|---|
APPEARANCE | WHITE TO OFF-WHITE POWDER |
SOLUBILITY | CLEAR COLORLESS TO FAINT YELLOW SOLUTION AT 50 MG/ML IN ETHANOL |
WATER BY KARL FISCHER | REPORT RESULT |
CARBON | 64.4 - 67.7% * |
NITROGEN | 12.5 - 13.2% * |
PURITY BY THIN LAYER CHROMATOGRAPHY | > OR = 98% |
NOTE | * ANHYDROUS BASIS |
DOCUMENT # | G9137/05/07/08/1 |