Product Name
MONOCLONAL ANTI-LC3 (APG8) antibody produced in mouse clone 166AT1234, IgG fraction of antiserum, buffered aqueous solution
Product Number
SAB1305552
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Appearance | Report result |
Concentration | Report result |
Purification | Protein G purified |
Western Blotting | 1:8000 |