Product Name
Anti-GPRC5D antibody produced in mouse purified immunoglobulin, buffered aqueous solution
Product Number
SAB1401754
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 - 5 ug/ml. Specific band of ~37.95 kDa using GPRC5D transfected 293T lysate. |
Flow Cytometry | Antibody concentration: 1 ug/ml using GPRC5D expressing 293 cells. |
Detection Antibody | Suitable as a detection antibody in sandwich ELISA assay. |