Product Name
Monoclonal Anti-LHX2, (N-terminal) antibody produced in mouse clone 1E6, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1402464
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
ELISA (Recombinant protein) | Antibody concentration: 1 ug/mL using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/mL. Specific band of ~34.47 kDa using immunogen protein lysate. |
Western Blotting (Transfected lysate) | Antibody concentration: 0.1 - 5 ug/mL. Specific band of ~44.4 kDa using LHX2 transfected 293T lysate. |