Product Name
Monoclonal Anti-WWTR1 antibody produced in mouse clone 1F1, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1402558
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 3 ug/mL using human placenta tissue. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/mL using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/mL. Specific band of ~37.11 kDa using immunogen protein lysate. |