Product Name
ANTI-PPARGC1A antibody produced in mouse clone 2F9, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1411922
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Appearance | colorless liquid |
Concentration | Report Result |
Storage/Stability | Report Result |
Storage Temperature | -20�C |
Species Reactivity | Report Result |
ELISA | Antibody concentration: 1 ug/ml using recombinant protein. |
Sandwich ELISA | Detection limit for recombinant GST tagged PPARGC1A is approximately 1ng/ml. |
Western Blotting (Recombinant Protein) | Antibody concentration: 1 ug/ml. Specific band of ~37.8 kDa using immunogen protein lysate. |
Western Blotting (Cell Lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~91 kDa using U-2 OS lysate. |
Western Blotting (Tissue lysate) | Not Verified |
Western Blotting (Transfected lysate) | Not Verified |
Immunohistochemistry | Not Verified |
Immunofluorescence | Not Verified |
Handling | Report Result |