Product Name
Anti-LYVE1 antibody produced in rabbit affinity isolated antibody
Product Number
SAB4300606
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Appearance | colorless liquid |
Concentration | 1.0 mg/ml |
ELISA | Not Determined |
Western Blotting | 1:500-1:1000 |
Immunohistochemistry | Not Determined |
Immunofluorescence | Not Determined |
Immunoprecipitation | Not Determined |
Flow Cytometry | Not Determined |
Chromatin Immunoprecipitation | Not Determined |