Product Name
Anti-Goat IgG (H+L), highly cross-adsorbed, CF™770 antibody produced in donkey ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Product Number
SAB4600374
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Appearance | blue solution |
Concentration | 1.8-2.2 mg/mL |
Purity | Negligible or no free dye detected |
Reactivity | goat IgG(H+L) |
Absorption Maximum | 770 +/- 5 nm |
Absorption Degree of Labeling (moles dye per moles protein) | 3-6 |