Product Name
Anti-Aquaporin-1 (AQP1) antibody produced in rabbit
Product Number
SAB5701511
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Concentration | Report result |
Western Blotting | 1:500 - 1:2000 |
Immunofluorescence | 1:50 - 1:200 |