Product Name
Amyloid β 26-35 human
Product Number
SCP0044
제품 브랜드
SIGMA
CAS Number
Molecular Weight
1003.22
테스트 | 규격 |
---|---|
Form | lyophilized powder |
Appearance | White to off-white |
Peptide Content | > or = to 68% |
Purity by HPLC | > or = to 95% |
테스트 | 규격 |
---|---|
Form | lyophilized powder |
Appearance | White to off-white |
Peptide Content | > or = to 68% |
Purity by HPLC | > or = to 95% |