Product Name
TRAIL, Soluble human recombinant, expressed in E. coli, His tagged, >98% (SDS-PAGE)
Product Number
SRP8024
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Form | Report result |
Color | Report result |
Concentration | Report result |
Purity by SDS-Page | ≥98% |
Endotoxin Level | <0.01 EU/µg purified protein (LAL test) |