Product Name
Monoclonal Anti-ACTN4 antibody produced in mouse clone 4D10, purified immunoglobulin, buffered aqueous solution
Product Number
WH0000081M1
제품 브랜드
SIGMA
CAS Number
Molecular Weight
테스트 | 규격 |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 - 5 ug/ml. Specific band of ~105 kDa using Hela NE lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 3 ug/ml using human testis tissue. |
Immunofluorescence (IF) | Antibody concentration: 20 ug/ml using HeLa cell. |
Sandwich ELISA | Detection limit for recombinant GST tagged ACTN4 is approximately 0.1 ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~38 kDa using immunogen protein lysate. |