Product Name
10-O-Deacetylpaclitaxel reference material
Product Number
29914
제품 브랜드
SUPELCO
CAS Number
78432-77-6
Molecular Weight
811.87
테스트 | 규격 |
---|---|
Assay (QNMR)* | Report certified value and uncertainty (acc. to ISO17025) |
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
EXPIRY PERIOD | 24 MONTHS |