뒤로
IKA
®
VRX basic Vibrax
®
orbital shaker attachments
Products
Manufacturer
직경(mm)
높이(mm)
포장 단위
제품 필터
Showing 1-2 of 2
비교
Product Number
설명
가격
Z404268
IKA
®
VRX basic Vibrax
®
orbital shaker attachments,
one-hand attachment, for shaking single, unfixed Erlenmeyer/shaking 250mL flasks, 1/cs
확장
가격 보기
Z404144
IKA
®
VRX basic Vibrax
®
orbital shaker attachments,
Erlenmeyer flask attachment, for Erlenmeyer flasks (2 flasks up to 500mL), 1/cs