뒤로
Kern EWJ series precision balances
Products
제품 카테고리
balances (6)
Manufacturer
직경(mm)
길이(mm)
칭량 용량(g)
포장 단위
제품 필터
Showing 1-6 of 6
비교
Product Number
설명
가격
Z742817
Kern EWJ series precision balances,
Kern EWJ 600-2M verified 965-216, weighing capacity 600 g, accuracy: 0.01 g, precision: ±0.03 g, AC/DC input 100 - 240 V AC, universal plug set
확장
가격 보기
Z742815
Kern EWJ series precision balances,
Kern EWJ 300-3, weighing capacity 300 g, accuracy: 0.001 g, precision: ±0.003 g
확장
가격 보기
Z742816
Kern EWJ series precision balances,
Kern EWJ 600-2M, weighing capacity 600 g, accuracy: 0.01 g, precision: ±0.03 g
확장
가격 보기
Z742819
Kern EWJ series precision balances,
Kern EWJ 6000-1M, weighing capacity 6000 g, accuracy: 0.1 g, precision: ±0.3 g
확장
가격 보기
Z742818
Kern EWJ series precision balances,
Kern EWJ 3000-2, weighing capacity 3000, accuracy: 0.01 g, precision: ±0.05 g
확장
가격 보기
Z742820
Kern EWJ series precision balances,
Kern EWJ 6000-1M verified 965-217, weighing capacity 6000 g, accuracy: 0.1 g, precision: ±0.3 g