뒤로
Kern FCB series bench scale
Products
제품 카테고리
balances (6)
Manufacturer
칭량 용량(g)
제품 필터
Showing 1-6 of 6
비교
Product Number
설명
가격
Z742912
Kern FCB series bench scale,
Kern FCB 30K1, weighing capacity 30000 g, accuracy: 1.0 g, precision: ±3.0 g
확장
가격 보기
Z742911
Kern FCB series bench scale,
Kern FCB 12K1, weighing capacity 12000 g, accuracy: 1.0 g, precision: ±3.0 g
확장
가격 보기
Z742907
Kern FCB series bench scale,
Kern FCB 3K0.1, weighing capacity 3000 g, accuracy: 0.1 g, precision: ±0.3 g
확장
가격 보기
Z742913
Kern FCB series bench scale,
Kern FCB 24K2, weighing capacity 24000 g, accuracy: 2.0 g, precision: ±6.0 g
확장
가격 보기
Z742909
Kern FCB series bench scale,
Kern FCB 6K0.5, weighing capacity 6000 g, accuracy: 0.5 g, precision: ±1.5 g
확장
가격 보기
Z742908
Kern FCB series bench scale,
Kern FCB 8K0.1, weighing capacity 8000 g, accuracy: 0.1 g, precision: ±0.3 g