Merck
Mini Diazald® apparatus joint: ST/NS 19/26 Clear-Seal joints, for generating 1 to 50 mmol of diazomethane
Mini Diazald® apparatus
Mini Diazald® apparatus joint: ST/NS 19/26 Clear-Seal joints, for generating 1 to 50 mmol of diazomethane