Sign Into View Organizational & Contract Pricing
About This Item
Volume:
40—1000 μL
Sterility:
non-sterile
Feature:
ATP free
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free
Packaging:
pack of 960 ea
Manufacturer/tradename:
BRAND 734654
material
polypropylene tip
sterility
non-sterile
feature
ATP free
PCR clean
RNase and DNase free
endotoxin free
filter
pyrogen free
packaging
pack of 960 ea
manufacturer/tradename
BRAND 734654
volume
40—1000 μL
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
For use with pipetting robots, tips are required that are matched to the robot′s mode of operation in terms of geometry and surface properties. BRAND® robotic pipette tips and robotic filter tips meet all requirements for use with the BRAND® Liquid Handling Station pipetting robot. The raw materials are free of DiHEMDA and Oleamid. Tips are manufactured at BRAND® under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used.
Features and Benefits
- BIO-CERT® LIQUID HANDLING QUALITY: Free of DNA, RNase, DNase, Pyrogens, PCR-Inhibitors and ATP
- For use with the BRAND® Liquid Handling Station pipetting robot
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service