Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ARHGEF6(9459)

General description

Rho guanine nucleotide exchange factor 6 (UniProt Q15052; also known as Alpha-Pix, COOL-2, PAK-interacting exchange factor alpha, Rac/Cdc42 guanine nucleotide exchange factor 6) is encoded by the ARHGEF6 (also known as COOL2, KIAA0006, MRX46, PIXA) gene (Gene ID 9459) in human. ARHGEF6 (alpha-Pix) belongs to a family of guanine nucleotide exchange factors (GEF) that activate the Rho family of small GTPases. ARHGEF6 acts as a Cdc42 and Rac1 GEF and forms a signling module with the adaptor GIT (GPCR kinase interactor), the GTPase effector PAK (p21 activated kinase), and the GTPase activator PIX (PAK-interacting exchange factor), facilitating the activation Rac and Cdc42 GTPases at specific subcellular locations. In contrast to the ubiquitious expression pattern of ARHGEF7 (beta-Pix), ARHGEF6 is only detected in highly motile cells, including immune cells, microvascular cells, and growing fetal neurons, where it plays an essential role in mediating cell spreading, directional migration, angiogenesis, and fetal neuron dendrite outgrowth. In addition, both ARHGEF6 and ARHGEF7 are shown to interact with the RING type E3 ubiquitin ligase c-Cbl. While ARHGEF6 is a substrate for c-Cbl-mediated proteasomal degradation, ARHGEF7 binds and antagonizes against c-Cbl-mediated EGFR proteasomal degradation. Elevated ARHGEF6 accumulation is found in human A172 and rat C6 glioma cells that do not express c-Cbl, resulting in enhanced cell migration and invasion.

Specificity

This polyclonal antibody targets an internal sequence C-terminal to the PH domain.

Immunogen

KLH-conjugated linear peptide corresponding to an internal sequence C-terminal to the PH domain.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a represenative lot detected ARHGEF6 in A431, HeLa, and NIH/3T3 cells.

Western Blotting Analysis: A 1:500 dilution from a represenative lot detected ARHGEF6 in 10 µg of human placenta tissue lysate as well as in 10 µg of A431, HeLa, HepG2, HUVEC, Jurkat, and PC3 cell lysates.

Research Category
Cell Structure
This rabbit polyclonal Anti-ARHGEF6 Antibody, Cat. No. ABT15 is validated for use in Immunocytochemistry and Western Blotting for the detection of Rho guanine nucleotide exchange factor 6.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected ARHGEF6 in 10 µg of HEK293 cell lysate.

Target description

~71 kDa observed. 87.50/70.79 kDa (human isoform 1/2), 87.05 kDa (mouse & rat) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service