Skip to Content
Merck
All Photos(3)

Key Documents

938114

Sigma-Aldrich

Dihydropinene

greener alternative

BioRenewable, reagent grade

Synonym(s):

Pinane, cis-pinane, 2,6,6-Trimethylbicyclo[3.1.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
5237941
MDL number:
UNSPSC Code:
41111500

grade

reagent grade

Quality Level

Assay

≥97%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

renewable carbon content

% (Confirmed)

color

colorless

refractive index

n/D 1.450-1.470

bp

167-172 °C

mp

<25 °C

density

0.857 g/cm3 at 20 °C
0.857 g/mL at 20 °C (lit.)

greener alternative category

SMILES string

CC1CC[C@@H]2C[C@H]1C2(C)C

InChI

1S/C10H18/c1-7-4-5-8-6-9(7)10(8,2)3/h7-9H,4-6H2,1-3H3/t7?,8-,9-/m1/s1

InChI key

XOKSLPVRUOBDEW-CFCGPWAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is BioRenewable and thus aligns with ″Use of Renewable Feedstocks″, and ″Safer Solvents and Auxiliaries″. Click here for more information. This reagent grade Dihydropinene can be used as an alternative to Hexane in certain applications. Made from renewable carbon (verified through C14 ASTM D6866-16 testing) and produced in an environmentally friendly way that reduces the carbon footprint of production and reliance on fossil fuels. Because it is not from petroleum, it is free from many non-renewable contaminants, such as benzene, phthalates, and other phenols, leading to a higher quality product.

Application

Dihydropinene can be used in the synthesis of quinazolones and was as in a variety of extraction applications.

Features and Benefits

Made from fully Bio-based material ASTM D6866 - Standard Test Methods for Determining the Bio-based Content Made from Renewable feedstock - wood tall oil An alternative to Hexane

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F

Flash Point(C)

42 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service