Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

FITC conjugate

Quality Level

antibody product type

primary antibodies

clone

MOPC-21, monoclonal

form

buffered aqueous solution

species reactivity

(This mouse IgG1 kappa monoclonal antibody with unknown specificity has been confirmed as a good negative control with human and rat species.)

concentration

1 mg/ml

technique(s)

flow cytometry: suitable

isotype

IgG1κ

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This mouse IgG1 kappa monoclonal antibody (clone MOPC-21) with unknown specificity has been confirmed as a good negative control with human and rat species, based on multiple testing on rat and human tissues.

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with fluorescein isothiocyanate (FITC) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service