Skip to Content
Merck
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2M22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 14 kDa
observed mol wt ~16 kDa

purified by

using Protein A

species reactivity

human, yeast

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

yeast ... HTA1(851811)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2M22 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Histone H2A phosphorylated on Serine 129.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids from the C-terminal region of yeast Histone H2A.1.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HeLa cells treated with UV.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-Histone H2A (Ser129) in lysate from HeLa cells treated with UV.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected p-Histone H2A in Yeast Extract.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected p-Histone H2A in HeLa cells serum-starved overnight -/+ Etoposide 20ug/mL for 3 hours.

Peptide Inhibition Assay: Target band detection in UV treated HeLa cells was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

A 1:1,000 dilution from a representative lot detected Histone H2A in HeLa treated with UV.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Histone H2A.1 (UniProt: P04911; also known as H2A) is encoded by the HTA1 (also known as H2A1, SPT11, YD9934.10, YDR225W) gene (Gene ID: 851811) in Saccharomyces cerevisiae (yeast). Histone H2A is one of the 5 main histones that are involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H2A is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histones are modified post-translationally by acetylation, phosphorylation, methylation, and ubiquitination and these modifications regulate DNA transcription, repair, recombination, and replication. Histone H2A.1 undergoes phosphorylation at Serine 129 that plays a crucial role in various cellular processes involved in DNA repair, genomic stability, and cell cycle regulation. Yeast histone H2A1 contains a [ST]-Q motif (aa 129-130) in the C-terminal region that is phosphorylated by the Mec1 and Tel1 kinases in response to DNA double strand breaks or DNA damage signals. It serves as a recognition mark for the recruitment of DNA repair factors and facilitates the repair of damaged DNA. Overall, Histone H2A.1 phosphorylation at serine 129 is a significant event in maintaining genomic integrity and cell homeostasis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service