Product Name
4-Chlorothiophenol 97%
Product Number
125237
Product Brand
ALDRICH
CAS Number
106-54-7
Molecular Weight
144.62
Test | Specification |
---|---|
APPEARANCE | WHITE TO OFF-WHITE SOLID, CRYSTALS OR |
CRYSTALLINE POWDER (OR COLORLESS LIQUID | |
AFTER MELTING) | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE AND STANDARD AS |
ILLUSTRATED ON PAGE 1178D OF EDITION I, | |
VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR | |
SPECTRA". | |
TITRATION | TYPICALLY 97% - 103% (WITH AGNO3 AFTER |
OXYGEN COMBUSTION) | |
GAS LIQUID | TYPICALLY >97% |
APPROVED JANUARY 13, 1999 JSB |