Product Name
(1-Chloroethyl)trimethylsilane technical, ≥90% (GC)
Product Number
23070
Product Brand
ALDRICH
CAS Number
7787-87-3
Molecular Weight
136.70
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Faint Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 90.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |