Product Name
Desyl bromide 97%
Product Number
258156
Product Brand
ALDRICH
CAS Number
1484-50-0
Molecular Weight
275.14
Test | Specification |
---|---|
APPEARANCE (COLOR) | Off White to Light Yellow |
APPEARANCE (FORM) | Powder |
TITRATION | 96.50 - 103.50 % |
TITRATION (METHOD) | BROMINE COMBUSTION |
PURITY (HPLC AREA %) | ≥ 96.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |