Product Name
2,6-Difluorobenzylamine 97%
Product Number
264393
Product Brand
ALDRICH
CAS Number
69385-30-4
Molecular Weight
143.13
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5%-103.5% (WITH HCLO4) |
GAS LIQUID | 96.5% (MINIMUM) |
APPROVED OCTOBER 23, 2007 RJM |