Product Name
Sodium phenoxide trihydrate 98%
Product Number
318191
Product Brand
ALDRICH
CAS Number
156150-40-2
Molecular Weight
170.14
Test | Specification |
---|---|
APPEARANCE | WHITE TO BEIGE TO PINK CRYSTALS OR POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 98.5%-101.5% (WITH HCLO4) CORRECTED FOR H2O |
AND/OR | |
GAS LIQUID | 98.5% (MINIMUM) |
REVIESED APRIL 19, 2006 JSB |