Product Name
2-Mercaptopyridine N-oxide sodium salt 95%
Product Number
329061
Product Brand
ALDRICH
CAS Number
3811-73-2
Molecular Weight
149.15
Test | Specification |
---|---|
APPEARANCE | WHITE TO LIGHT YELLOW TO TAN POWDER |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 94.0% - 106.0% (WITH HCLO4) |
SOLUBILITY | 5% IN H2O; CLEAR, COLORLESS TO LIGHT YELLOW |
SOLUTION | |
REVISED MARCH 1, 2007 RJM |