Product Name
1,3-Diaminopropane-N,N,N′,N′-tetraacetic acid ≥99.0% (KT)
Product Number
33267
Product Brand
ALDRICH
CAS Number
1939-36-2
Molecular Weight
306.27
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Faint Beige |
APPEARANCE (FORM) | Powder |
TITRATION (KT) ZNSO4 0.1M | 99.0 - 101.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |