Product Name
α-Bromo-α-methyl-γ-butyrolactone 96%
Product Number
393568
Product Brand
ALDRICH
CAS Number
33693-67-3
Molecular Weight
179.01
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 95.5% (MINIMUM) |
APPROVED SEPTEMBER 30, 2008 RJM |